Thermo Scientific Chemicals

9-Phenyl-2,3,7-trihydroxy-6-fluorone, Thermo Scientific Chemicals

Catalog number: A11495.06
5 g, Each
Thermo Scientific Chemicals

9-Phenyl-2,3,7-trihydroxy-6-fluorone, Thermo Scientific Chemicals

Catalog number: A11495.06
5 g, Each
Quantity
Catalog number: A11495.06
also known as A11495-06
Price (USD)
Price: 55.40
Online price: 47.65
Your price:
Quantity
-

Chemical Identifiers

CAS
975-17-7
IUPAC Name
2,6,7-trihydroxy-9-phenyl-3H-xanthen-3-one
Molecular Formula
C19H12O5
InChI Key
YDCFOUBAMGLLKA-UHFFFAOYSA-N
SMILES
OC1=CC2=C(C3=CC=CC=C3)C3=CC(O)=C(O)C=C3OC2=CC1=O
Appearance (Color)
Orange to brown
Water Content (Karl Fischer Titration)
≤1%
Form
Powder
lambda-max (UV-Vis)
550 ? 5 nm in aq. NaOH
Sulfated Ash
≤2.5%

Description

9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).

Solubility
Soluble in chloroform.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents. Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text