4-n-Pentylphenol, 98%
4-n-Pentylphenol, 98%
4-n-Pentylphenol, 98%
Thermo Scientific Chemicals

4-n-Pentylphenol, 98%

CAS: 14938-35-3 | C11H16O | 164.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11514.14
also known as A11514-14
Price (EUR)
155,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
155,00
Each
Chemical Identifiers
CAS14938-35-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to pale brown
FormLiquid
Assay (GC)≥97.0%
Refractive Index1.5120-1.5160 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only