Malonic acid, 99%
Malonic acid, 99%
Malonic acid, 99%
Thermo Scientific Chemicals

Malonic acid, 99%

CAS: 141-82-2 | C3H4O4 | 104.061 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11526.0E
also known as A11526-0E
Price (TWD)
-
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS141-82-2
IUPAC Namepropanedioic acid
Molecular FormulaC3H4O4
InChI KeyOFOBLEOULBTSOW-UHFFFAOYSA-N
SMILESOC(=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or granules
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. sourced material)
Assay from Supplier's CofA≥98.5% (U.S. sourced material)
CommentMaterial Sourced in the U.S. and in other countries
View more
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.

Solubility
Soluble in cold water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Knoevenagel condensation with aldehydes yields substituted acrylic (e.g. cinnamic) acids; review: Org. React., 15, 204 (1967). For an example of the Doebner modification, using pyridine as solvent/base, see: Org. Synth. Coll., 3, 425 (1955); a catalytic amount of piperidine often gives superior results; see, e.g.: Org. Synth. Coll., 4, 327 (1963).
  2. Bew, S. P.; Stephenson, R.; Rouden, J.; Ashford, P. A.; Bourane, M.; Charvet, A.; Dalstein, V. M. D.; Jauseau, R.; Gipson, G. D. H.; Lozano, L. A. M. Bioinspired, Base- and Metal-Free, Mild Decarboxylative Aldol Activation of Malonic Acid Half Thioesters Under Phase-Transfer Reaction Conditions. Adv. Synth. Catal. 2015, 357 (6), 1245-1257.
  3. Nakamura, S.; Sano, M.; Toda, A.; Nakane, D.; Masuda, H. Organocatalytic Enantioselective Decarboxylative Reaction of Malonic Acid Half Thioesters with Cyclic N-Sulfonyl Ketimines by Using N-Heteroarenesulfonyl Cinchona Alkaloid Amides. Chem. Eur. J. 2015, 21 (10), 3929-3932.