Methyl p-tolyl sulfone, 99%
Methyl p-tolyl sulfone, 99%
Methyl p-tolyl sulfone, 99%
Thermo Scientific Chemicals

Methyl p-tolyl sulfone, 99%

CAS: 3185-99-7 | C8H10O2S | 170.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11531.14
also known as A11531-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3185-99-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder or lumps
Melting Point (clear melt)86.0-90.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only