2-Fluorobenzyl chloride, 98+%
2-Fluorobenzyl chloride, 98+%
2-Fluorobenzyl chloride, 98+%
Thermo Scientific Chemicals

2-Fluorobenzyl chloride, 98+%

CAS: 345-35-7 | C7H6ClF | 144.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11566.22
also known as A11566-22
Price (TWD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS345-35-7
IUPAC Name1-(chloromethyl)-2-fluorobenzene
Molecular FormulaC7H6ClF
InChI KeyMOBRMRJUKNQBMY-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.5135-1.5175 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only