Diethyl 2-(2-cyanoethyl)malonate, 98+%
Diethyl 2-(2-cyanoethyl)malonate, 98+%
Diethyl 2-(2-cyanoethyl)malonate, 98+%
Diethyl 2-(2-cyanoethyl)malonate, 98+%
Thermo Scientific Chemicals

Diethyl 2-(2-cyanoethyl)malonate, 98+%

CAS: 17216-62-5 | C10H15NO4 | 213.233 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
25 g
50 g
產品號碼 A11567.14
亦稱為 A11567-14
價格 (TWD)
-
申請報價
Quantity:
25 g
Request bulk or custom format
化學識別
CAS17216-62-5
IUPAC Name1,3-diethyl 2-(2-cyanoethyl)propanedioate
Molecular FormulaC10H15NO4
InChI KeyYJJLOESDBPRZIP-UHFFFAOYSA-N
SMILESCCOC(=O)C(CCC#N)C(=O)OCC
檢視更多
規格Specification Sheet規格表
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4345-1.4385 @ 20?C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only