1,2,4-Triazole, 99%
1,2,4-Triazole, 99%
1,2,4-Triazole, 99%
Thermo Scientific Chemicals

1,2,4-Triazole, 99%

CAS: 288-88-0 | C2H3N3 | 69.067 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A11597.36
または、製品番号A11597-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS288-88-0
IUPAC Name1H-1,2,4-triazole
Molecular FormulaC2H3N3
InChI KeyNSPMIYGKQJPBQR-UHFFFAOYSA-N
SMILESN1C=NC=N1
さらに表示
Formcrystalline powder or flakes
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)>98.5%
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Triazole is widely used as an intermediate in the manufacture of pharmaceuticals. It acts as an anti-scaling agent and antifungal compound. It is widely used in antifungals such as fluconazole anditraconazole.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Feldman, J.; Vo, G. D.; McLaren, C. D.; Gehret, T. C.; Park, K.; Meth, J. S.; Marshall, W. J.; Buriak, J.; Bryman, L. M.; Dobbs, K. D.; Scholz, T. H.; Zane, S. G. Highly Quantum Efficient Phosphorescent Sky Blue Emitters Based on Diastereomeric Iridium(III) Complexes of Atropisomeric 5-Aryl-4H-1,2,4-triazole Ligands. Organometallics 2015, 34 (15), 3665-3669.
  2. Bennion, J. C.; McBain, A.; Son, S. F.; Matzger, A. J. Design and Synthesis of a Series of Nitrogen-Rich Energetic Cocrystals of 5,5'-Dinitro-2H,2H'-3,3'-bi-1,2,4-triazole (DNBT). Cryst. Growth Des. 2015, 15 (5), 2545-2549.