Aluminum acetate, basic hydrate
Aluminum acetate, basic hydrate
Aluminum acetate, basic hydrate
Thermo Scientific Chemicals

Aluminum acetate, basic hydrate

CAS: 142-03-0 | C4H7AlO5 | 162.08 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A11620.22
または、製品番号A11620-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS142-03-0
IUPAC Name(acetyloxy)(hydroxy)alumanyl acetate
Molecular FormulaC4H7AlO5
InChI KeyHQQUTGFAWJNQIP-UHFFFAOYSA-K
SMILESCC(=O)O[Al](O)OC(C)=O
さらに表示
Assay from Suppliers CofA≥98.0% (U.S. specification)
CommentAl2O3 content ≥ 28.0% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Elemental AnalysisAluminium Al: ≥ 11.28% to ≤15.77 (non-U.S. specification)
Insoluble material≤0.05% (non-U.S. specification)
さらに表示
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.

Solubility
Insoluble in water

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Al-Ghamdi, A. A.; Al-Hartomy, O. A.; El Okr, M.; Nawar, A. M.; El-Gazzar, S.; El-Tantawy, F.; Yakuphanoglu, F. Semiconducting properties of Al doped ZnO thin films. Spectrochim. Acta, Part A 2014, 131, 512-517.
  2. Shin, H. U.; Stefaniak, A. B.; Stojilovic, N.; Chase, G. G. Comparative dissolution of electrospun Al2O3 nanofibres in artificial human lung fluids. Environ. Sci.: Nano 2015, 2 (3), 251-261.