1-Dimethylamino-2-nitroethylene, 98%
1-Dimethylamino-2-nitroethylene, 98%
1-Dimethylamino-2-nitroethylene, 98%
Thermo Scientific Chemicals

1-Dimethylamino-2-nitroethylene, 98%

CAS: 1190-92-7 | C4H8N2O2 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A11637.06
also known as A11637-06
Price (EUR)
119,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
119,00
Each
Chemical Identifiers
CAS1190-92-7
IUPAC Namedimethyl(2-nitroethenyl)amine
Molecular FormulaC4H8N2O2
InChI KeyJKOVQYWMFZTKMX-UHFFFAOYSA-N
SMILESCN(C)C=C[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to dark brown to brown
Assay (GC)≥97.5%
Melting Point (clear melt)99.0-107.0°C
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only