2-Bromo-2-nitropropane-1,3-diol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-2-nitropropane-1,3-diol, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number A11639.36
also known as A11639-36
Price (USD)/ Each
355.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
418.00 
Save 62.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
355.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
418.00 
Save 62.35 (15%)
Add to cart
2-Bromo-2-nitropropane-1,3-diol, 98+%, Thermo Scientific Chemicals
Catalog numberA11639.36
Price (USD)/ Each
355.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
418.00 
Save 62.35 (15%)
-
Add to cart
Chemical Identifiers
CAS52-51-7
IUPAC Name2-bromo-2-nitropropane-1,3-diol
Molecular FormulaC3H6BrNO4
InChI KeyLVDKZNITIUWNER-UHFFFAOYSA-N
SMILESOCC(Br)(CO)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥98.0 to ≤102.0%
Assay (GC)≥98.0%
Identification (FTIR)Conforms
View more
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.

Solubility
Soluble in water(280g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Frances J. Storrs, M.D.; Diana E. Bell, R.D. Allergic contact dermatitis to 2-bromo-2-nitropropane-1,3-diol in a hydrophilic ointment. Journal of the American Academy of Dermatology. 1983, 8, (2), 157-170.