2-Octanone, 98%
2-Octanone, 98%
2-Octanone, 98%
Thermo Scientific Chemicals

2-Octanone, 98%

CAS: 111-13-7 | C8H16O | 128.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
5000 mL
Catalog number A11643.AU
also known as A11643-AU
Price (EUR)
84,00
Each
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (EUR)
84,00
Each
Add to cart
Chemical Identifiers
CAS111-13-7
IUPAC Nameoctan-2-one
Molecular FormulaC8H16O
InChI KeyZPVFWPFBNIEHGJ-UHFFFAOYSA-N
SMILESCCCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4130-1.4170 @ 20?C
Assay (GC)≥97.5%
2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only
M Krogh; H Grefslie; K E Rasmussen. Solvent-modified solid-phase microextraction for the determination of diazepam in human plasma samples by capillary gas chromatography. Journal of chromatography. B, Biomedical sciences and applications.1997, 689 (2), 357-364.