Thermo Scientific Chemicals

Decanal, 96%, Thermo Scientific Chemicals

Catalog number: A11656.AP
500 mL, Each
Thermo Scientific Chemicals

Decanal, 96%, Thermo Scientific Chemicals

Catalog number: A11656.AP
500 mL, Each
Quantity
Catalog number: A11656.AP
also known as A11656-AP
Price (USD)
Price: 156.00
Online price: 135.65
Your price:
Quantity
-

Chemical Identifiers

CAS
112-31-2
IUPAC Name
decanal
Molecular Formula
C10H20O
InChI Key
KSMVZQYAVGTKIV-UHFFFAOYSA-N
SMILES
CCCCCCCCCC=O
Assay (GC)
≥95.0%
Appearance (Color)
Clear colorless
Free acid (titration)
≤2%
Form
Liquid
Refractive Index
1.4250-1.4310 @ 20?C

Description

Decanal is used in fragrances and flavoring and it is an important component in citrus along with octanal, citral, and sinensal. It is also used in Air Care Products, Laundry and Dishwashing Products, Personal Care Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decanal is used in fragrances and flavoring and it is an important component in citrus along with octanal, citral, and sinensal. It is also used in Air Care Products, Laundry and Dishwashing Products, Personal Care Products.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text