2-Chloro-2-methylbutane, 95%
2-Chloro-2-methylbutane, 95%
2-Chloro-2-methylbutane, 95%
Thermo Scientific Chemicals

2-Chloro-2-methylbutane, 95%

CAS: 594-36-5 | C5H11Cl | 106.593 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11703.14
also known as A11703-14
Price (EUR)
102,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
102,00
Each
Chemical Identifiers
CAS594-36-5
IUPAC Name2-chloro-2-methylbutane
Molecular FormulaC5H11Cl
InChI KeyCRNIHJHMEQZAAS-UHFFFAOYSA-N
SMILESCCC(C)(C)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4015-1.4075 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Avoid extremes of temperature and direct sunlight.
RUO – Research Use Only
Lídia M.P.C. Albuquerque; M.Luisa C.J. Moita; M.N. Simoes; Raquel M.C. Goncalves. The enthalpies of solution of 2-chloro-2-methylpropane, 2-chloro-2-methylbutane and 3-chloro-3-methylpentane in mono- and dialcohols at 298.15 K. Thermochimica acta. 1996, 275 67-73.