5-Chloro-1,3-dimethyl-1H-pyrazole, 98%
5-Chloro-1,3-dimethyl-1H-pyrazole, 98%
5-Chloro-1,3-dimethyl-1H-pyrazole, 98%
5-Chloro-1,3-dimethyl-1H-pyrazole, 98%
Thermo Scientific Chemicals

5-Chloro-1,3-dimethyl-1H-pyrazole, 98%

CAS: 54454-10-3 | C5H7ClN2 | 130.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11730.14
also known as A11730-14
Price (EUR)
317,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
317,00
Each
Add to cart
Chemical Identifiers
CAS54454-10-3
IUPAC Name5-chloro-1,3-dimethyl-1H-pyrazole
Molecular FormulaC5H7ClN2
InChI KeyDDUSLFAWARYAPR-UHFFFAOYSA-N
SMILESCN1N=C(C)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4845-1.4885 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only