1-Chloro-4-phenylbutane, 97%
1-Chloro-4-phenylbutane, 97%
1-Chloro-4-phenylbutane, 97%
Thermo Scientific Chemicals

1-Chloro-4-phenylbutane, 97%

CAS: 4830-93-7 | C10H13Cl | 168.664 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A11753.06
also known as A11753-06
Price (EUR)
116,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
116,00
Each
Chemical Identifiers
CAS4830-93-7
IUPAC Name(4-chlorobutyl)benzene
Molecular FormulaC10H13Cl
InChI KeyFLLZCZIHURYEQP-UHFFFAOYSA-N
SMILESClCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5160-1.5210 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only