Thermo Scientific Chemicals

Sodium metaborate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A11803.36
500 g, Each
Thermo Scientific Chemicals

Sodium metaborate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A11803.36
500 g, Each
Quantity
Catalog number: A11803.36
also known as A11803-36
Price (USD)
Price: 36.80
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10555-76-7
IUPAC Name
sodium tetrahydrate oxoborinate
Molecular Formula
BH8NaO6
InChI Key
JAKYJVJWXKRTSJ-UHFFFAOYSA-N
SMILES
O.O.O.O.[Na+].[O-]B=O
Form
Crystals or powder or crystalline powder or fused solid
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text