Thermo Scientific Chemicals

N-Methylformanilide, 99%, Thermo Scientific Chemicals

Catalog number: A11829.36
500 g, Each
Thermo Scientific Chemicals

N-Methylformanilide, 99%, Thermo Scientific Chemicals

Catalog number: A11829.36
500 g, Each
Quantity
Catalog number: A11829.36
also known as A11829-36
Price (USD)
Price: 143.00
Online price: 122.65
Your price:
Quantity
-

Chemical Identifiers

CAS
93-61-8
IUPAC Name
N-methyl-N-phenylformamide
Molecular Formula
C8H9NO
InChI Key
JIKUXBYRTXDNIY-UHFFFAOYSA-N
SMILES
CN(C=O)C1=CC=CC=C1
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.5590-1.5630 @ 20?C

Description

N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.

Solubility
Immisible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text