4,4'-Stilbenedicarboxylic acid, 96%
4,4'-Stilbenedicarboxylic acid, 96%
4,4'-Stilbenedicarboxylic acid, 96%
Thermo Scientific Chemicals

4,4'-Stilbenedicarboxylic acid, 96%

CAS: 100-31-2 | C16H12O4 | 268.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A11839.06
also known as A11839-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS100-31-2
IUPAC Name4-[(1E)-2-(4-carboxyphenyl)ethenyl]benzoic acid
Molecular FormulaC16H12O4
InChI KeySBBQDUFLZGOASY-OWOJBTEDSA-N
SMILESOC(=O)C1=CC=C(C=CC2=CC=C(C=C2)C(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormPowder
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only