Thermo Scientific Chemicals

2-Acetyl-5-chlorothiophene, 99%, Thermo Scientific Chemicals

10 g, Each
Thermo Scientific Chemicals

2-Acetyl-5-chlorothiophene, 99%, Thermo Scientific Chemicals

10 g, Each
Quantity
Catalog number: A11850.09
also known as A11850-09
Price (USD)
Price: 38.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
6310-09-4
IUPAC Name
1-(5-chlorothiophen-2-yl)ethan-1-one
Molecular Formula
C6H5ClOS
InChI Key
HTZGPEHWQCRXGZ-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(Cl)S1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (GC)
≥98.5%
Melting Point (clear melt)
44.0-48.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only