(±)-3-Butyn-2-ol, 98%
(±)-3-Butyn-2-ol, 98%
(±)-3-Butyn-2-ol, 98%
Thermo Scientific Chemicals

(±)-3-Butyn-2-ol, 98%

CAS: 2028-63-9 | C4H6O | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
100 g
Catalog number A11881.06
also known as A11881-06
Price (EUR)
34,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
34,50
Each
Chemical Identifiers
CAS2028-63-9
IUPAC Namebut-3-yn-2-ol
Molecular FormulaC4H6O
InChI KeyGKPOMITUDGXOSB-UHFFFAOYNA-N
SMILESCC(O)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation0.0 ± 0.1? (neat)
Appearance (Color)Clear colorless to yellow
Refractive Index1.4235-1.4275 @ 20?C
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only