4-Bromo-1,2-difluorobenzene, 98+%
4-Bromo-1,2-difluorobenzene, 98+%
4-Bromo-1,2-difluorobenzene, 98+%
Thermo Scientific Chemicals

4-Bromo-1,2-difluorobenzene, 98+%

CAS: 348-61-8 | C6H3BrF2 | 192.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11934.14
also known as A11934-14
Price (EUR)
92,10
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
92,10
Each
Chemical Identifiers
CAS348-61-8
IUPAC Name4-bromo-1,2-difluorobenzene
Molecular FormulaC6H3BrF2
InChI KeyYMQPKONILWWJQG-UHFFFAOYSA-N
SMILESFC1=CC=C(Br)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or very pale green
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5025-1.5065 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only