3-Bromothiophene-2-carboxylic acid, 97%
3-Bromothiophene-2-carboxylic acid, 97%
3-Bromothiophene-2-carboxylic acid, 97%
3-Bromothiophene-2-carboxylic acid, 97%
Thermo Scientific Chemicals

3-Bromothiophene-2-carboxylic acid, 97%

CAS: 7311-64-0 | C5H3BrO2S | 207.041 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 A11935.06
亦稱為 A11935-06
價格 (TWD)
-
申請報價
Quantity:
5 g
Request bulk or custom format
化學識別
CAS7311-64-0
IUPAC Name3-bromothiophene-2-carboxylate
Molecular FormulaC5H2BrO2S
InChI KeyVQQLWBUTTMNMFT-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=C(Br)C=CS1
檢視更多
規格Specification Sheet規格表
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)192.0-201.0?C
Appearance (Color)White to cream to pale brown
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only