2'-Bromoacetophenone, 99%
2'-Bromoacetophenone, 99%
2'-Bromoacetophenone, 99%
Thermo Scientific Chemicals

2'-Bromoacetophenone, 99%

CAS: 2142-69-0 | C8H7BrO | 199.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11982.09
also known as A11982-09
Price (TWD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS2142-69-0
IUPAC Name1-(2-bromophenyl)ethan-1-one
Molecular FormulaC8H7BrO
InChI KeyPIMNFNXBTGPCIL-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5670-1.5700 @ 20?C
FormLiquid
2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Adolfo Lopez Torres.; Eunice Yanez Barrientos.; Katarzyna Wrobel.; Kazimierz Wrobel. Selective Derivatization of Cytosine and Methylcytosine Moieties with 2-Bromoacetophenone for Submicrogram DNA Methylation Analysis by Reversed Phase HPLC with Spectrofluorimetric Detection.Anal. Chem. 2011, 83 (20),7999-8005.
  2. S. Ramalingam.;P. Anbusrinivasan.; S. Periandy. FT-IR and FT-Raman spectral investigation, computed IR intensity and Raman activity analysis and frequency estimation analysis on 4-chloro-2-bromoacetophenone using HF and DFT calculations.Spectrochim. Acta, Part A. 2011, 78 (2),826-834.