1,3-Dibenzoyloxybenzene, 98%, Thermo Scientific Chemicals
1,3-Dibenzoyloxybenzene, 98%, Thermo Scientific Chemicals
1,3-Dibenzoyloxybenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dibenzoyloxybenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A11983.22
also known as A11983-22
100 g
Catalog number A11983.22
also known as A11983-22
Price (USD)
298.65
Special offer
Online exclusive
Ends: 30-Jun-2026
351.00
Save 52.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
298.65
Special offer
Online exclusive
Ends: 30-Jun-2026
351.00
Save 52.35 (15%)
Each
Chemical Identifiers
CAS94-01-9
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)114.0-120.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only