Thermo Scientific Chemicals

4-Ethoxybenzaldehyde, 97+%, Thermo Scientific Chemicals

Catalog number: A11987.22
100 g, Each
Thermo Scientific Chemicals

4-Ethoxybenzaldehyde, 97+%, Thermo Scientific Chemicals

Catalog number: A11987.22
100 g, Each
Quantity
Catalog number: A11987.22
also known as A11987-22
Price (USD)
Price: 95.70
Online price: 83.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10031-82-0
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.0%
Free acid (titration)
≤1.5%
Identification (FTIR)
Conforms
Refractive Index
1.5560-1.5610 @ 20?C

Description

4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents. Air sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text