Thermo Scientific Chemicals

Palladium, 10% on carbon, Type 487, dry, Thermo Scientific Chemicals

Catalog number: A12012.04
2 g, Each
Thermo Scientific Chemicals

Palladium, 10% on carbon, Type 487, dry, Thermo Scientific Chemicals

Catalog number: A12012.04
2 g, Each
Quantity
Catalog number: A12012.04
also known as A12012-04
Price (USD)
Price: 121.00
Online price: 102.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7440-05-3
IUPAC Name
palladium
Molecular Formula
Pd
InChI Key
KDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES
[Pd]
Appearance (Color)
Black
Form
Powder
Assay from Suppliers CofA
Palladium content 9-11% (dry wt. basis)
Water Content
≤2.5%

Description

It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.

Solubility
Insoluble in water and organic solvents.

Notes
Palladium on carbon should always be handled under an inert atmosphere (preferably argon), and reaction vessels should be flushed with inert gas before the catalyst is added.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text