4-Chlorothiobenzamide, 97%
4-Chlorothiobenzamide, 97%
4-Chlorothiobenzamide, 97%
Thermo Scientific Chemicals

4-Chlorothiobenzamide, 97%

CAS: 2521-24-6 | C7H6ClNS | 171.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
100 g
Catalog number A12021.14
also known as A12021-14
Price (EUR)
70,50
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
70,50
Each
Chemical Identifiers
CAS2521-24-6
IUPAC Name4-chlorobenzene-1-carbothioamide
Molecular FormulaC7H6ClNS
InChI KeyOKPUICCJRDBRJT-UHFFFAOYSA-N
SMILESNC(=S)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Melting Point (clear melt)121.5-130.5?C
Assay (HPLC)≥97.5%
FormPowder
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Qi-Dong You.; Hou-Yuan Zhou.; Qi-Zhou Wang.; Xing-Han Lei. Preparation Of N-Unsubstituted Thioamides By A Modified Willgerodt-Kindler Reaction. New Journal for Organic Synthesis. 1991, 23 (4),435-438.
  2. Steven J. Coats .; Jeffrey S. Link.; Dennis J. Hlasta. Alternative Solvents for Elevated-Temperature Solid-Phase Parallel Synthesis. Application to Thionation of Amides.Org. Lett. 2003, 5 (5),721-724.