Chloramine-T trihydrate, 98%
Chloramine-T trihydrate, 98%
Chloramine-T trihydrate, 98%
Thermo Scientific Chemicals

Chloramine-T trihydrate, 98%

CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol
数量:
250 g
1000 g
5000 g
製品番号(カタログ番号) A12044.0I
価格(JPY)
-
数量:
5000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7080-50-4
IUPAC Namesodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
Molecular FormulaC7H13ClNNaO5S
InChI KeyRNROOTWAJBXJBD-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl
さらに表示
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Assay (Iodometric Titration)≥97.5 to ≤102.5%
Appearance (Color)White
Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

Solubility
Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.

Notes
Incompatible with strong oxidizing agents, acids and ammonia.
RUO – Research Use Only

General References:

  1. Mal, S.; Prathap, K. J.; Smith, S. C.; Umarye, J. D. Facile one pot synthesis of 8-chloro-[1,2,4]triazolo[4,3-a]pyrazines via oxidative cyclisation using chloramine T. Tetrahedron Lett. 2015, 56 (22), 2896-2901.
  2. Dzandzi, J. P. K.; Vera, D. R. B.; Genady, A. R.; Albu, S. A.; Eltringham-Smith, L. J.; Capretta, A.; Sheffield, W.P.; Valliant, J. F. Fluorous Analogue of Chloramine-T: Preparation, X-ray Structure Determination, and Use as an Oxidant for Radioiodination and s-Tetrazine Synthesis. J. Org. Chem. 2015, 80 (14), 7117-7125.