4-Azaxanthone, 97%
4-Azaxanthone, 97%
4-Azaxanthone, 97%
Thermo Scientific Chemicals

4-Azaxanthone, 97%

CAS: 6537-46-8 | C12H7NO2
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12045.03
also known as A12045-03
Price (EUR)
40,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
40,00
Each
Add to cart
Chemical Identifiers
CAS6537-46-8
IUPAC Name10H-chromeno[3,2-b]pyridin-10-one
Molecular FormulaC12H7NO2
InChI KeyYAUODDQCNOBEDJ-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2OC2=CC=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting Point180-189°C
Assay (HPLC)≥96.0%
FormPowder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only