Thermo Scientific Chemicals

Dimethoxymethane, 98%, Thermo Scientific Chemicals

Catalog number: A12055.AP
500 mL, Each
Thermo Scientific Chemicals

Dimethoxymethane, 98%, Thermo Scientific Chemicals

Catalog number: A12055.AP
500 mL, Each
Quantity
Catalog number: A12055.AP
also known as A12055-AP
Price (USD)
Price: 34.90
Online price: 29.65
Your price:
Quantity
-

Chemical Identifiers

CAS
109-87-5
IUPAC Name
dimethoxymethane
Molecular Formula
C3H8O2
InChI Key
NKDDWNXOKDWJAK-UHFFFAOYSA-N
SMILES
COCOC
Appearance (Color)
Clear, colourless
Assay (GC)
>97.5%
Form
liquid
Water Content (Karl Fischer Titration)
<1.0%
Refractive Index
1.3530-1.3560

Description

Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:

Solubility
Soluble in water 20°C 330g/L.

Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text