2',4'-Dihydroxyacetophenone, 98%
2',4'-Dihydroxyacetophenone, 98%
2',4'-Dihydroxyacetophenone, 98%
Thermo Scientific Chemicals

2',4'-Dihydroxyacetophenone, 98%

CAS: 89-84-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12066.30
also known as A12066-30
Price (EUR)
134,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
134,00
Each
Chemical Identifiers
CAS89-84-9
IUPAC Name1-(2,4-dihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O3
InChI KeySULYEHHGGXARJS-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Melting Point (clear melt)142.0-148.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to pink
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only