Thermo Scientific Chemicals

4-Bromo-2-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals

5g, Each
Thermo Scientific Chemicals

4-Bromo-2-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals

5g, Each
Quantity
Catalog number: A12146.06
also known as A12146-06
Price (USD)
Price: 50.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
445-02-3
IUPAC Name
4-bromo-2-(trifluoromethyl)aniline
Molecular Formula
C7H5BrF3N
InChI Key
PHXGKHTWEOPCEW-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C=C1C(F)(F)F
Refractive Index
1.5310-1.5350 @ 20?C
Form
Liquid
Appearance (Color)
Clear pale yellow
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only