Thermo Scientific™

p-Toluenesulfonamide, 98+%, Thermo Scientific Chemicals

Catalog number: A12156.0E
2500 g, Each
Thermo Scientific™

p-Toluenesulfonamide, 98+%, Thermo Scientific Chemicals

Catalog number: A12156.0E
2500 g, Each
Quantity
Catalog number: A12156.0E
also known as A12156-0E
Price (JPY)

Chemical Identifiers

CAS
70-55-3
IUPAC Name
4-methylbenzene-1-sulfonamide
Molecular Formula
C7H9NO2S
InChI Key
LMYRWZFENFIFIT-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(N)(=O)=O
Appearance (Color)
White
Assay (GC)
≥98.0%
Solution Test
5% in Ethanol: clear
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

p-Toluene sulfonamide was used to prepare the precursor required for synthesis of ethyl 6-phenyl-1-tosyl-1,2,5,6-tetrahydropyridine-3-carboxylate. It act as a derivative of ammonia activated to alkylation by alkyl halides is exemplified by the synthesis of N-tosyl-2,3-dihydroisoindole from o-xylylene dibromide. It is a precursor of N-tosyl imines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluene sulfonamide was used to prepare the precursor required for synthesis of ethyl 6-phenyl-1-tosyl-1,2,5,6-tetrahydropyridine-3-carboxylate. It act as a derivative of ammonia activated to alkylation by alkyl halides is exemplified by the synthesis of N-tosyl-2,3-dihydroisoindole from o-xylylene dibromide. It is a precursor of N-tosyl imines.

Solubility
Soluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text