Thermo Scientific Chemicals

4-Methylmorpholine, 99%, Thermo Scientific Chemicals

Catalog number: A12158.AE
100 mL, Each
Thermo Scientific Chemicals

4-Methylmorpholine, 99%, Thermo Scientific Chemicals

Catalog number: A12158.AE
100 mL, Each
Quantity
Catalog number: A12158.AE
also known as A12158-AE
Price (USD)
Price: 34.90
Online price: 29.65
Your price:
Quantity
-

Chemical Identifiers

CAS
109-02-4
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4340-1.4370 @ 20?C

Description

4-Methylmorpholine is used as a solvent for dyes, resins, waxes and pharmaceuticals. It acts as a crosslinker in the preparation of polyurethane foams, elastomers and adhesives. It is used as a precursor to prepare N-methylmorpholine N-oxide and morpholinium cation based ionic liquids. It is utilized as corrosion inhibitors and anti-scaling agents in industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylmorpholine is used as a solvent for dyes, resins, waxes and pharmaceuticals. It acts as a crosslinker in the preparation of polyurethane foams, elastomers and adhesives. It is used as a precursor to prepare N-methylmorpholine N-oxide and morpholinium cation based ionic liquids. It is utilized as corrosion inhibitors and anti-scaling agents in industries.

Solubility
Miscible with water.

Notes
Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text