Sulfur trioxide-pyridine complex, 98%, active SO3 ca 48-50%
Sulfur trioxide-pyridine complex, 98%, active SO3 ca 48-50%
Sulfur trioxide-pyridine complex, 98%, active SO3 ca 48-50%
Thermo Scientific Chemicals

Sulfur trioxide-pyridine complex, 98%, active SO3 ca 48-50%

CAS: 26412-87-3 | C5H5NO3S | 159.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
1000 g
Catalog number A12202.09
also known as A12202-09
Price (EUR)
27,50
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
27,50
Each
Chemical Identifiers
CAS26412-87-3
IUPAC Namepyridine; sulfonylideneoxidane
Molecular FormulaC5H5NO3S
InChI KeyUDYFLDICVHJSOY-UHFFFAOYSA-N
SMILESO=S(=O)=O.C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pale yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
FormPowder
Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.

Solubility
Soluble in polar organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Converts alcohols to monoalkyl sulfates, utilized in carbohydrate protection: Carbohydr. Res., 127, 211, 131, C8 (1984); J. Org. Chem., 38, 3510 (1973). The pyridinium sulfates of allylic or benzylic alcohols can be deoxygenated in high yields by in situ LiAlH4 reduction in THF: J. Org. Chem., 34, 3667 (1969); Tetrahedron Lett., 4650 (1972); J. Am. Chem. Soc., 93, 7016 (1971).
  2. Sulfonates indole at the 3-position: Tetrahedron, 29, 669 (1973).
  3. The combination with Dimethyl sulfoxide, A13280, is a mild oxidant for alcohols to carbonyl compounds (Parikh-Doering Reagent): J. Am. Chem. Soc., 89, 5505 (1967), avoiding the very low temperatures required for the Swern and similar systems (see Oxalyl chloride, A18012). With chiral alcohols, less racemization occurs at the ɑ-carbon than with Collins' reagent or pyridinium dichromate: Tetrahedron Lett., 23, 807 (1982); J. Org. Chem., 46, 4799 (1981). Nicolaou utilized the system at 0°C to convert an epoxy alcohol to the corresponding aldehyde in the synthesis of hemibrevitoxin B: J. Am. Chem. Soc., 115, 3558 (1993).
  4. In combination with NaI, reduces ɑ-halo ketones to ketones: Synthesis, 59 (1979), and sulfoxides to sulfides: Synthesis, 984 (1979).
  5. Lim, D. K.; Wylie, R. G.; Langer, R.; Kohane, D. S. Selective binding of C-6 OH sulfated hyaluronic acid to the angiogenic isoform of VEGF165. Biomaterials 2016, 77, 130-138.
  6. Sarbova, V.; Koschella, A.; Cheng, F.; Kelly, S. M.; Heinze, T. Studies on the sulfation of cellulose alpha-lipoate and ability of the sulfated product to stabilize colloidal suspensions of gold nanoparticles. Carbohydr. Polym. 2015, 124, 117-123.