2-氨基-3'-硝基苯乙酮 盐酸盐, 98%
2-氨基-3'-硝基苯乙酮 盐酸盐, 98%
2-氨基-3'-硝基苯乙酮 盐酸盐, 98%
Thermo Scientific Chemicals

2-氨基-3'-硝基苯乙酮 盐酸盐, 98%

CAS: 36765-84-1 | C8H9N2O3 | 181.17 g/mol
Have Questions?
更改视图buttonViewtableView
数量:
1 g
5 g
货号 A12205.03
又称 A12205-03
价格(TWD)
-
数量:
1 g
请求批量或定制报价
化学标识符
CAS36765-84-1
规格Specification Sheet规格表
Appearance (Color)White to cream to pale brown
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Melting Point (clear melt)205-212?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only