2-Amino-3'-nitroacetophenone hydrochloride, 98%
2-Amino-3'-nitroacetophenone hydrochloride, 98%
2-Amino-3'-nitroacetophenone hydrochloride, 98%
Thermo Scientific Chemicals

2-Amino-3'-nitroacetophenone hydrochloride, 98%

CAS: 36765-84-1 | C8H9N2O3 | 181.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12205.06
also known as A12205-06
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS36765-84-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Melting Point (clear melt)205-212?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only