Amplex™ Red Phospholipase D Assay Kit
Invitrogen™

Amplex™ Red Phospholipase D Assay Kit

Detect phospholipase D activity levels as low as 10U/mL

Have Questions?
Catalog NumberQuantity
A12219500 Assays
Catalog number A12219
Price (USD)
494.00
Each
Quantity:
500 Assays
Price (USD)
494.00
Each

The Amplex™ Red Phospholipase D Assay Kit provides a sensitive and simplemethod to detect Phospholipase D (PLD) activity using a fluorescence microplate reader or fluorometer.

  • Detect phospholipase D activity levels as low as 10 U/mL
  • Format allows for multiple time point measurements
  • Designed for minimal autofluorescence interference

PLD activity is monitored indirectly using 10-acetyl-3,7-dihydroxyphenoxazine (Amplex™ Red reagent), a sensitive fluorogenic probe for hydrogen peroxide. PLD converts the phosphatidylcholine (lecithin) to choline, which is then oxidized by choline oxidase to betaine and hydrogen peroxide. In the presence of horseradish peroxidase, huydrogen peroxide reacts with the Amplex™ Red reagent in a 1:1 stoichiometric ratio to generate the highly fluorescent product resorufin.

Because resorufin has absorption and fluorescence emission maxima of approximately 571 nm and 585 nm, respectively, there is little interference from autofluorescence in most biological samples.

Use Amplex™ Red Assays for a Broad Range of Investigations

A wide variety of validated Amplex™ Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex™ UltraRed Reagent (Cat. No. A36006), a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex™ Red/UltraRed Stop Reagent (Cat. No. A33855). The Amplex™ Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours. Custom assay design and packaging are also available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeAmplex Red
Emission Wavelength585 nm
Excitation Wavelength571 nm
Quantity500 Assays
Shipping ConditionRoom Temperature
Substrate PropertiesLipid-Based Substrate, Chemical Substrate
Substrate TypePhospholipase Substrate
Target EnzymePhospholipase
For Use With (Application)Enzyme and Metabolic Activity
For Use With (Equipment)Microplate Reader
Product LineAmplex
Product TypePhospholipase Assay
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

I'm using an Amplex Red kit, the reagent changes color to pink almost immediately in my own Krebs-Ringer buffer but not in HBSS. Why is this?

The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can Amplex Red Assays be performed using cell lysates?

This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.

The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (17)

Citations & References
Abstract
Phospholipase D1 production of phosphatidic acid at the plasma membrane promotes exocytosis of large dense-core granules at a late stage.
Authors:Zeniou-Meyer M, Zabari N, Ashery U, Chasserot-Golaz S, Haeberlé AM, Demais V, Bailly Y, Gottfried I, Nakanishi H, Neiman AM, Du G, Frohman MA, Bader MF, Vitale N
Journal:J Biol Chem
PubMed ID:17540765
'Substantial efforts have recently been made to demonstrate the importance of lipids and lipid-modifying enzymes in various membrane trafficking processes, including calcium-regulated exocytosis of hormones and neurotransmitters. Among bioactive lipids, phosphatidic acid (PA) is an attractive candidate to promote membrane fusion through its ability to change membrane topology. To date, ... More
Cholesterol distribution in the Golgi complex of DITNC1 astrocytes is differentially altered by fresh and aged amyloid beta-peptide-(1-42).
Authors:Igbavboa U, Pidcock JM, Johnson LN, Malo TM, Studniski AE, Yu S, Sun GY, Wood WG
Journal:J Biol Chem
PubMed ID:12584199
'The Golgi complex plays an important role in cholesterol trafficking in cells, and amyloid beta-peptides (Abetas) alter cholesterol trafficking. The hypothesis was tested that fresh and aged Abeta-(1-42) would differentially modify Golgi cholesterol content in DINTC1 astrocytes and that the effects of Abeta-(1-42) would be associated with the region of ... More
The antisignaling agent SC-alpha alpha delta 9, 4-(benzyl-(2-[(2,5-diphenyloxazole-4-carbonyl)amino]ethyl)carbamoyl)- 2-decanoylaminobutyric acid, is a structurally unique phospholipid analogue with phospholipase C inhibitory activity.
Authors:Vogt A, Pestell KE, Day BW, Lazo JS, Wipf P,
Journal:Mol Cancer Ther
PubMed ID:12481409
'Phospholipids and lipid second messengers mediate mitogenic signal transduction and oncogenesis, but there have been few successful examples of small molecules that affect biologically important phospholipid metabolism. Here we investigated the actions of a previously described antitumor agent, 4-(benzyl-(2-[(2,5-diphenyloxazole-4-carbonyl)amino]ethyl)carbamoyl)- 2-decanoylaminobutyric acid (SC-alpha alpha delta 9), which has antisignaling properties, on ... More
Interfacial sensing by alveolar type II cells: a new concept in lung physiology?
Authors:Ravasio A, Hobi N, Bertocchi C, Jesacher A, Dietl P, Haller T,
Journal:Am J Physiol Cell Physiol
PubMed ID:21270294
'Alveolar type II (AT II) cells are in close contact with an air-liquid interface (I(AL)). This contact may be of considerable physiological relevance; however, no data exist to provide a satisfying description of this specific microenvironment. This is mainly due to the experimental difficulty to manipulate and analyze cell-air contacts ... More
Loss of the ceramide transfer protein augments EGF receptor signaling in breast cancer.
Authors:Heering J, Weis N, Holeiter M, Neugart F, Staebler A, Fehm TN, Bischoff A, Schiller J, Duss S, Schmid S, Korte T, Herrmann A, Olayioye MA,
Journal:Cancer Res
PubMed ID:22472120
'Triple-negative breast cancers (TNBC) are especially refractory to treatment due to their negative hormone receptor and ErbB2/HER2 status. Therefore, the identification of cancer-associated deregulated signaling pathways is necessary to develop improved targeted therapies. Here, we show that expression of the ceramide transfer protein CERT is reduced in TNBCs. CERT transfers ... More