2-Phenoxyethylamine, 98%
2-Phenoxyethylamine, 98%
2-Phenoxyethylamine, 98%
Thermo Scientific Chemicals

2-Phenoxyethylamine, 98%

CAS: 1758-46-9 | C8H11NO | 137.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12225.14
also known as A12225-14
Price (EUR)
473,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
473,00
Each
Chemical Identifiers
CAS1758-46-9
IUPAC Name2-phenoxyethan-1-aminium
Molecular FormulaC8H12NO
InChI KeyIMLAIXAZMVDRGA-UHFFFAOYSA-O
SMILES[NH3+]CCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSpecification differs for U.S. and non-U.S. material where indicated
Refractive Index1.5360-1.5400 @ 20°C (non-U.S. specification)
FormLiquid
Infrared spectrumConforms (U.S. specification)
Appearance (Color)Clear colorless to yellow
View more
Organic intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only