Thermo Scientific Chemicals

2-Fluorobiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: A12227.14
25 g, Each
Thermo Scientific Chemicals

2-Fluorobiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: A12227.14
25 g, Each
Quantity
Catalog number: A12227.14
also known as A12227-14
Price (USD)
Price: 295.00
Online price: 257.65
Your price:
Quantity
-

Chemical Identifiers

CAS
321-60-8
IUPAC Name
2-fluoro-1,1'-biphenyl
Molecular Formula
C12H9F
InChI Key
KLECYOQFQXJYBC-UHFFFAOYSA-N
SMILES
FC1=CC=CC=C1C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream
Melting Point (clear melt)
71.0-77.0?C
Assay (GC)
≥97.5%

Description

2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text