Thermo Scientific Chemicals

Benzophenone hydrazone, 98+%, Thermo Scientific Chemicals

Catalog number: A12266.18
50 g, Each
Thermo Scientific Chemicals

Benzophenone hydrazone, 98+%, Thermo Scientific Chemicals

Catalog number: A12266.18
50 g, Each
Quantity
Catalog number: A12266.18
also known as A12266-18
Price (USD)
Price: 28.40
Online price: 23.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5350-57-2
IUPAC Name
(diphenylmethylidene)hydrazine
Molecular Formula
C13H12N2
InChI Key
QYCSNMDOZNUZIT-UHFFFAOYSA-N
SMILES
NN=C(C1=CC=CC=C1)C1=CC=CC=C1
Form
crystals or crystalline powder
Appearance (Color)
White to yellow or pale brown
Assay (GC)
>98.0%

Description

An important compound in organic photochemistry and perfumery as well as in organic synthesis. Used in the antibiotics synthesis of 6-APA and 7-ACA of the carboxyl protecting groups and other organic compounds. Used as organic pigment and medical intermediate. It is used as a photoinitiator of UV-curing applications in inks, adhesive, coatings and optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An important compound in organic photochemistry and perfumery as well as in organic synthesis. Used in the antibiotics synthesis of 6-APA and 7-ACA of the carboxyl protecting groups and other organic compounds. Used as organic pigment and medical intermediate. It is used as a photoinitiator of UV-curing applications in inks, adhesive, coatings and optical fiber.

Solubility
Soluble in ether, benzene, chloroform and other organic solvents.

Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances. Keep refrigerated. (Store below 4 C/39 F.)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text