delta-Valerolactam, 98%
delta-Valerolactam, 98%
delta-Valerolactam, 98%
delta-Valerolactam, 98%
Thermo Scientific Chemicals

delta-Valerolactam, 98%

CAS: 675-20-7 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12271.22
also known as A12271-22
Price (TWD)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS675-20-7
IUPAC Namepiperidin-2-one
Molecular FormulaC5H9NO
InChI KeyXUWHAWMETYGRKB-UHFFFAOYSA-N
SMILESO=C1CCCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or brown
FormCrystalline or fused solid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)<2.0%
View more
delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Naoki Toyooka; Yasuko Yoshida; Yasuhito Yotsui and Takefumi Momose. 2-Piperidone Type of Chiral Building Block for 3-Piperidinol Alkaloid Synthesis. J. Org. Chem. 1999, 64 (13), 4914-4919.
  2. Tetsuji Kametani; Terumi Higa; Masuo Koizumi; Masataka Ihara; Keiichiro Fukumoto. Iminoketene cycloaddition. 2. Total syntheses of arborine, glycosminine, and rutecarpine by condensation of iminoketene with amides. J. Am. Chem. Soc. 1977, 99 (7), 2306-2309.
  3. Starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition: J. Org. Chem., 58, 5041 (1993).
  4. For reaction as a heterodienophile in cycloaddition with Danishefsky's diene, see: Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato) europium(III), 33541.