Thermo Scientific Chemicals

3-Chloropropiophenone, 96%, Thermo Scientific Chemicals

Catalog number: A12290.22
100 g, Each
Thermo Scientific Chemicals

3-Chloropropiophenone, 96%, Thermo Scientific Chemicals

Catalog number: A12290.22
100 g, Each
Quantity
Catalog number: A12290.22
also known as A12290-22
Price (USD)
Price: 262.00
Online price: 252.65
Your price:
Quantity
-

Chemical Identifiers

CAS
936-59-4
IUPAC Name
3-chloro-1-phenylpropan-1-one
Molecular Formula
C9H9ClO
InChI Key
KTJRGPZVSKWRTJ-UHFFFAOYSA-N
SMILES
ClCCC(=O)C1=CC=CC=C1
Appearance (Color)
White to pale cream to pale brown
Melting Point (clear melt)
42.0-52.0?C
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥95.0%

Description

3-Chloropropiophenone was used in the asymmetric reduction of (S)-3-chloro-1-phenylpropanol using preheated Candida utilis cells immobilized in calcium alginate gel beads. It was also used in the synthesis of (R)-3-chloro-1-phenyl-1-propanol via asymmetric reduction using in-situ generated oxazaborolidine catalyst derived from (S)-α,α-diphenylprolinol. It is a precursor of phenyl vinyl ketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropiophenone was used in the asymmetric reduction of (S)-3-chloro-1-phenylpropanol using preheated Candida utilis cells immobilized in calcium alginate gel beads. It was also used in the synthesis of (R)-3-chloro-1-phenyl-1-propanol via asymmetric reduction using in-situ generated oxazaborolidine catalyst derived from (S)-α,α-diphenylprolinol. It is a precursor of phenyl vinyl ketone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text