Thermo Scientific Chemicals

4-Nitrophenyl phosphate disodium salt hexahydrate, 96%, Thermo Scientific Chemicals

Catalog number: A12310.14
25 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl phosphate disodium salt hexahydrate, 96%, Thermo Scientific Chemicals

Catalog number: A12310.14
25 g, Each
Quantity
Catalog number: A12310.14
Price (USD)
Price: 174.00
Online price: 147.65
Your price:
Quantity
-

Chemical Identifiers

CAS
333338-18-4
IUPAC Name
disodium hexahydrate 4-nitrophenyl phosphate
Molecular Formula
C6H16NNa2O12P
InChI Key
KAKKHKRHCKCAGH-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
Form
Crystalline powder or flakes
Assay (HPLC)
≥95.0% (U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
White to pale yellow
Assay (Non-aqueous acid-base Titration)
≥95.0 to ≤105.0% (non-U.S. specification)

Description

Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.

Solubility
Miscible with water. Immiscible with ethanol and acetone.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text