Thermo Scientific Chemicals

3-Methylcatechol, 97%, Thermo Scientific Chemicals

Catalog number: A12324.22
100 g, Each
Thermo Scientific Chemicals

3-Methylcatechol, 97%, Thermo Scientific Chemicals

Catalog number: A12324.22
100 g, Each
Quantity
Catalog number: A12324.22
also known as A12324-22
Price (USD)
Price: 364.00
Online price: 315.65
Your price:
Quantity
-

Chemical Identifiers

CAS
488-17-5
IUPAC Name
3-methylbenzene-1,2-diol
Molecular Formula
C7H8O2
InChI Key
PGSWEKYNAOWQDF-UHFFFAOYSA-N
SMILES
CC1=CC=CC(O)=C1O
Appearance (Color)
White to cream to brown
Assay (GC)
≥96.0%
Melting Point (clear melt)
60.0-69.0?C
Form
Crystals or powder or crystalline powder or fused solid or lumps

Description

Used in organic synthesis. Used for the synthesis of antibacterial agent, antioxidants, As a polymer inhibitor, stabilizer, flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in organic synthesis. Used for the synthesis of antibacterial agent, antioxidants, As a polymer inhibitor, stabilizer, flavors.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text