Thermo Scientific Chemicals

Sulfadiazine, 98%, Thermo Scientific Chemicals

Catalog number: A12370.30
250 g, Each
Thermo Scientific Chemicals

Sulfadiazine, 98%, Thermo Scientific Chemicals

Catalog number: A12370.30
250 g, Each
Quantity
Catalog number: A12370.30
also known as A12370-30
Price (TWD)

Chemical Identifiers

CAS
68-35-9
IUPAC Name
4-amino-N-(pyrimidin-2-yl)benzene-1-sulfonamide
Molecular Formula
C10H10N4O2S
InChI Key
SEEPANYCNGTZFQ-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CC=N1
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥97.5% (¹H-NMR) (U.S. specification)
Appearance (Color)
White
Assay (HPLC)
≥97.5% (U.K. specification)
Proton NMR
Conforms to structure (U.S. specification)

Description

Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text