2,4-Dichlorothiophenol, 97%
2,4-Dichlorothiophenol, 97%
2,4-Dichlorothiophenol, 97%
Thermo Scientific Chemicals

2,4-Dichlorothiophenol, 97%

CAS: 1122-41-4 | C6H4Cl2S | 179.058 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12384.03
also known as A12384-03
Price (EUR)
26,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
26,70
Each
Chemical Identifiers
CAS1122-41-4
IUPAC Name(2,4-dichlorophenyl)sulfanide
Molecular FormulaC6H3Cl2S
InChI KeyFGBVJFREPSJSNG-UHFFFAOYSA-M
SMILES[S-]C1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.6150-1.6200 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only