Thermo Scientific Chemicals

Phosphoenolpyruvic acid mono(cyclohexylammonium) salt, 98%, Thermo Scientific Chemicals

Catalog number: A12385.14
25 g, Each
Thermo Scientific Chemicals

Phosphoenolpyruvic acid mono(cyclohexylammonium) salt, 98%, Thermo Scientific Chemicals

Catalog number: A12385.14
25 g, Each
Quantity
Catalog number: A12385.14
also known as A12385-14
Price (USD)
Price: 1,271.00
Online price: 1,099.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10526-80-4
IUPAC Name
2-(phosphonooxy)prop-2-enoic acid; cyclohexanamine
Molecular Formula
C9H18NO6P
InChI Key
VHFCNZDHPABZJO-UHFFFAOYSA-N
SMILES
NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to yellow
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text