4'-Phenoxyacetophenone, 98+%
4'-Phenoxyacetophenone, 98+%
4'-Phenoxyacetophenone, 98+%
4'-Phenoxyacetophenone, 98+%
Thermo Scientific Chemicals

4'-Phenoxyacetophenone, 98+%

CAS: 5031-78-7 | C14H12O2 | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12392.22
also known as A12392-22
Price (EUR)
283,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
283,00
Each
Add to cart
Chemical Identifiers
CAS5031-78-7
IUPAC Name1-(4-phenoxyphenyl)ethan-1-one
Molecular FormulaC14H12O2
InChI KeyDJNIFZYQFLFGDT-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder or flakes
Melting Point47.5-53.5?C
Appearance (Color)White to pale yellow or pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only