Piperidine, 99%
Thermo Scientific Chemicals

Piperidine, 99%

Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 A12442.AP
價格 (TWD)
-
Quantity:
500 mL
Request bulk or custom format
化學識別
CAS110-89-4
IUPAC Namepiperidine
Molecular FormulaC5H11N
InChI KeyNQRYJNQNLNOLGT-UHFFFAOYSA-N
SMILESC1CCNCC1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5 (GC: U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
檢視更多
Extensively finds applications as a base, a nucleophile and a solvent. Used for the production of dipiperidinyl dithiuram tetrasulfide, which is used as a rubber vulcanization accelerator. It is commonly used in solid phase peptide synthesis (F-MOC Chemistry) and in chemical degradation reactions (sequencing of DNA). Used in making rubber, curing epoxy resins and in fine chemicals, and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Piperidine, 99% 500mL Plastic bottle CAS No. 110-89-4